(S)-(-)-Limonen 96%
Glass bottle, 500 mL
Leverandør: Thermo Fisher Scientific
(S)-(-)-Limonen 96%
Katalog # ACRO203735000
Leverandør: Thermo Fisher Scientific
CAS nummer: 5989-54-8
Gå til:
Product Details & Documents
Synonymer: (S)-(-)-p-Mentha-1,8-dien, (S)-(-)-Dipenten, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexen, L(-)-Limonen
- CAS nummer:5989-54-8
- Formel:C₁₀H₁₆
- MDL nummer:MFCD00001558
- Molecular Weight:136.24 g/mol
- Merck Index:13,05515
- Kogende Pt:175 °C (1013 hPa)
- Smeltende Pt:104…84 °C
- Tæthed:0,842 g/cm³ (20 °C)
- Flash Pt:48 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:227-815-6
- FN:2052
- ADR:3,III
Documents
ACRO20373
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4690 to 1.4760 (20°C, 589 nm)
- GC:≥95 %
- Specific optical rotation:(20°C, 589 nm) (neat) ≤ -60°
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL
- Cat. No.:ACRO203735000



