Bariumnitrat 99+% for analysis
Plastic bottle, 2,5 kg
Leverandør: Thermo Fisher Scientific
Bariumnitrat 99+% for analysis
Katalog # ACRO203150025
Leverandør: Thermo Fisher Scientific
CAS nummer: 10022-31-8
Gå til:
Product Details & Documents
- CAS nummer:10022-31-8
- Formel:Ba(NO₃)₂
- MDL nummer:MFCD00003442
- Molecular Weight:261.34 g/mol
- Merck Index:13,00986
- Smeltende Pt:592 °C
- Tæthed:~ 3,23 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:233-020-5
- FN:1446
- ADR:5.1,II
Documents
ACRO20315
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Crystals
- Titration Complexometric:≥99.0 %
- pH:5 to 8 (5 % at 20°C)
- Chloride (Cl):≤10 ppm
- Calcium (Ca):≤50 ppm
- Iron (Fe):≤5 ppm
- Strontium (Sr):≤1000 ppm
- Heavy metals (as Pb):≤10 ppm
Specifikationer
- Pakning:Plastic bottle
- Pakn:2,5 kg



