Orders LinkContactUsLinkComponent
Hydroxylammoniumchlorid 97%
Hydroxylammoniumchlorid 97%
Plastic drum, 5 kg
Catalog # ACRO170360050
Corrosive
Toxic
Health
Environment
Hydroxylammoniumchlorid 97%
Catalog # ACRO170360050
CAS Number:  5470-11-1
Hydroxylammoniumchlorid 97%
Hydroxylammoniumchlorid 97%
Plastic drum, 5 kg
CAS Number:  5470-11-1
Price
5.850,00 kr
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Hydroxylamine HCl, Hydroxylamine hydrochloride
  • CAS Number:5470-11-1
  • Formula:NH₂OH·HCl
  • MDL Number:MFCD00051089
  • Molecular Weight:69.49 g/mol
  • Merck Index:13,04853
  • Boiling Pt:306 °C (1013 hPa)
  • Melting Pt:155…158 °C (dec.)
  • Density:1,67 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:226-798-2
  • UN:2923
  • ADR:8,III
Documents
Specifications
Specification Test Results
  • Appearance:
    White to light yellow Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    151 to 157 °C (Dec)
  • Titration with NaOH:
    ≥96.0 %
Specifications
  • Pakning:
    Plastic drum
  • Pakn:
    5 kg