Tannin 95%
Plastic drum, 5 kg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Gallotannin, Tannin
- CAS nummer:1401-55-4
- Formel:C₇₆H₅₂O₄₆
- MDL nummer:MFCD00066397
- Molecular Weight:1701.2 g/mol
- Merck Index:12,09221
- Smeltende Pt:218 °C (dec.)
- Tæthed:Min, 1 g/cm³ (20 °C)
- Flash Pt:198 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:215-753-2
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Yellow to brown Powder
- Infrared spectrum:Conforms
- Assay:≥94 % (on dry substance)
- Loss on drying:≤12.0 %
- Sulfated ash:≤1.0 %
Specifikationer
- Pakning:Plastic drum
- Pakn:5 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



