Ammoniumnitrat 99+% for analysis
Plastic bottle, 2,5 kg
Leverandør: Thermo Fisher Scientific
Ammoniumnitrat 99+% for analysis
Katalog # ACRO205860025
Leverandør: Thermo Fisher Scientific
CAS nummer: 6484-52-2
Gå til:
Product Details & Documents
- CAS nummer:6484-52-2
- Formel:NH₄NO₃
- MDL nummer:MFCD00011425
- Molecular Weight:80.04 g/mol
- Merck Index:13,00538
- Kogende Pt:210 °C (760 mmHg)
- Smeltende Pt:169 °C
- Tæthed:1,72 g/cm³
- Opbevaringstemperatur:Stue temperatur
- EINECS:229-347-8
- FN:1942
- ADR:5.1,III
Documents
ACRO20586
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Adhering crystals
- Residue after ignition:≤0.01 % (as SO4)
- pH:4.5 to 6 (5 % at 20°C)
- Titration with NaOH:≥99.0 %
- Chloride (Cl):≤5 ppm
- Nitrite (NO2):≤5 ppm
- Sulfate (SO4):≤20 ppm
- Iron (Fe):≤2 ppm
- Heavy metals (as Pb):≤5 ppm
Specifikationer
- Pakning:Plastic bottle
- Pakn:2,5 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



