Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Benzophenon 99+%, pure

Plastic drum, 5 kg
Katalog # ACRO219680050
Sundhed
Leverandør:   Thermo Fisher Scientific
Benzophenon 99+%, pure
Katalog # ACRO219680050
CAS nummer:  119-61-9
Benzophenon 99+%, pure
Benzophenon 99+%, pure

Plastic drum, 5 kg

CAS nummer:  119-61-9
Price
2.750,00 kr
Log ind for tilgængelighed
1 * 5 kg
Product Details & Documents
Synonymer:  Diphenylmethanone, Diphenyl ketone
  • CAS nummer:119-61-9
  • Formel:C₁₃H₁₀O
  • MDL nummer:MFCD00003076
  • Molecular Weight:182.22 g/mol
  • Merck Index:13,01099
  • Kogende Pt:305,4 °C (1013 hPa)
  • Smeltende Pt:47…49 °C
  • Tæthed:1,085 g/cm³ (20 °C)
  • Flash Pt:143 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-337-6
  • ADR:9,III
Specifikationer

Specifikationstestresultater

  • Appearance:
    White to off-white Crystalline powder or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    47 to 49 °C
  • GC:
    ≥99.0 %

Specifikationer

  • Pakning:
    Plastic drum
  • Pakn:
    5 kg

Specifikationer