Benzophenon 99+%, pure
Glass bottle, 1 kg
Leverandør: Thermo Fisher Scientific
Benzophenon 99+%, pure
Katalog # ACRO219680010
Leverandør: Thermo Fisher Scientific
CAS nummer: 119-61-9
Gå til:
Product Details & Documents
Synonymer: Diphenylmethanone, Diphenyl ketone
- CAS nummer:119-61-9
- Formel:C₁₃H₁₀O
- MDL nummer:MFCD00003076
- Molecular Weight:182.22 g/mol
- Merck Index:13,01099
- Kogende Pt:305,4 °C (1013 hPa)
- Smeltende Pt:47…49 °C
- Tæthed:1,085 g/cm³ (20 °C)
- Flash Pt:143 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-337-6
- ADR:9,III
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to off-white Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:47 to 49 °C
- GC:≥99.0 %
Specifikationer
- Pakning:Glass bottle
- Pakn:1 kg



