Hydroquinon 99.5%
Plastic drum, 5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,4-Dihydroxybenzen, Benzen-1,4-diol
- CAS Number:123-31-9
- Formula:C₆H₄(OH)₂
- MDL Number:MFCD00002339
- Molecular Weight:110.11 g/mol
- Merck Index:13,04833
- Boiling Pt:285…287 °C (760 mmHg)
- Melting Pt:170…174 °C
- Density:1,332 g/cm³ (20 °C)
- Flash Pt:165 °C
- Storage Temperature:Stue temperatur
- EINECS:204-617-8
- UN:3077
- ADR:9,III
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals or needles
- Infrared spectrum:Conforms
- Melting point:171 to 174 °C
- Water:≤0.6 % (K.F.)
- Sulfated ash:≤0.05 %
- HPLC:≥99.4 %
Specifications
- Pakning:Plastic drum
- Pakn:5 kg



