Hydroquinon 99.5%
Plastic bottle, 1 kg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: 1,4-Dihydroxybenzen, Benzen-1,4-diol
- CAS nummer:123-31-9
- Formel:C₆H₄(OH)₂
- MDL nummer:MFCD00002339
- Molecular Weight:110.11 g/mol
- Merck Index:13,04833
- Kogende Pt:285…287 °C (760 mmHg)
- Smeltende Pt:170…174 °C
- Tæthed:1,332 g/cm³ (20 °C)
- Flash Pt:165 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-617-8
- FN:3077
- ADR:9,III
Specifikationer
Specifikationstestresultater
- Appearance:White to off-white Crystalline powder or crystals or needles
- Infrared spectrum:Conforms
- Melting point:171 to 174 °C
- Water:≤0.6 % (K.F.)
- Sulfated ash:≤0.05 %
- HPLC:≥99.4 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 kg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



