Lithiumbromid, anhydrous 99+% for analysis
Plastic bottle, 10 kg
- :7550-35-8
- :LiBr
- :MFCD00011077
- :86.84 g/mol
- :13,05548
- :1265 °C (760 mmHg)
- :550 °C
- :3,46 g/cm³
- :Stue temperatur
- :231-439-8
Documents
- Appearance:White Powder
- Titration Argentometric:≥99.0 %
- Loss on drying:≤0.5 % (400°C)
- pH:6 to 12 (4 % at 20°C)
- Chloride (Cl):≤1500 ppm
- Carbonate (CO3):≤300 ppm
- Sulfate (SO4):≤200 ppm
- Calcium (Ca):≤50 ppm
- Copper (Cu):≤3 ppm
- Iron (Fe):≤10 ppm
- Magnesium (Mg):≤20 ppm
- Potassium (K):≤300 ppm
- Sodium (Na):≤400 ppm
- Insoluble matter:≤0.005 % (in water)
- Pakning:Plastic bottle
- Pakn:10 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



