Lithiumbromid, anhydrous 99+% for analysis
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Lithiumbromid, anhydrous 99+% for analysis
Catalog # ACRO199870025
Supplier: Thermo Fisher Scientific
CAS Number: 7550-35-8
Jump to:
Product Details & Documents
- CAS Number:7550-35-8
- Formula:LiBr
- MDL Number:MFCD00011077
- Molecular Weight:86.84 g/mol
- Merck Index:13,05548
- Boiling Pt:1265 °C (760 mmHg)
- Melting Pt:550 °C
- Density:3,46 g/cm³
- Storage Temperature:Stue temperatur
- EINECS:231-439-8
Documents
Specifications
Specification Test Results
- Appearance:White Powder
- Titration Argentometric:≥99.0 %
- Loss on drying:≤0.5 % (400°C)
- pH:6 to 12 (4 % at 20°C)
- Chloride (Cl):≤1500 ppm
- Carbonate (CO3):≤300 ppm
- Sulfate (SO4):≤200 ppm
- Calcium (Ca):≤50 ppm
- Copper (Cu):≤3 ppm
- Iron (Fe):≤10 ppm
- Magnesium (Mg):≤20 ppm
- Potassium (K):≤300 ppm
- Sodium (Na):≤400 ppm
- Insoluble matter:≤0.005 % (in water)
Specifications
- Pakning:Plastic bottle
- Pakn:2,5 kg



