Thiomorpholin 97%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:123-90-0
- Formula:C₄H₉NS
- MDL Number:MFCD00005974
- Molecular Weight:103.19 g/mol
- Density:1,088 g/cm³
- Flash Pt:63 °C
- Storage Temperature:Stue temperatur
- UN:3267
- ADR:8,III
Documents
ACRO18797
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5370 to 1.5390 (20°C, 589 nm)
- GC:≥96.0 %
Specifications
- Pakning:Glass bottle
- Pakn:25 g



