ACES 99%, extra pure
Glass bottle, 100 g
Leverandør: Thermo Fisher Scientific
ACES 99%, extra pure
Katalog # ACRO228001000
Leverandør: Thermo Fisher Scientific
CAS nummer: 7365-82-4
Gå til:
Product Details & Documents
Synonymer: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid
- CAS nummer:7365-82-4
- Formel:H₂NCOCH₂NHCH₂CH₂SO₃H
- MDL nummer:MFCD00008030
- Molecular Weight:182.2 g/mol
- Merck Index:13,00037
- Smeltende Pt:270…295 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:230-908-4
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤1 % (K.F.)
- UV:(5 % in water)
- pH:3.6 to 4.4 (1 % solution)
- Titration with NaOH:≥98.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:100 g



