Chloramphenicol 98%
Glass bottle, 500 g
: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
- :56-75-7
- :Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
- :MFCD00078159
- :323.13 g/mol
- :13,02087
- :645 °C (1013 hPa)
- :149…153 °C
- :1,49 g/cm³ (20 °C)
- :Stue temperatur
- :200-287-4
Documents
ACRO22792
SDS
- Appearance:White to yellowish-white to greyish-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:149 to 153 °C
- Loss on drying:≤0.5 % (105 °C)
- UV spectrophotometry:98.0 to 102.0 % (on dried substance)
- Specific optical rotation:+18.5° to +20.5° (20 °C, 589 nm) (c=6, ethanol)
- Pakning:Glass bottle
- Pakn:500 g



