Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Chloramphenicol 98%

Glass bottle, 100 g
Catalog # ACRO227921000
Health
Chloramphenicol 98%
Catalog # ACRO227921000
CAS Number:  56-75-7
Chloramphenicol 98%
Chloramphenicol 98%

Glass bottle, 100 g

CAS Number:  56-75-7
Price
1.360,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
  • CAS Number:56-75-7
  • Formula:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
  • MDL Number:MFCD00078159
  • Molecular Weight:323.13 g/mol
  • Merck Index:13,02087
  • Boiling Pt:645 °C (1013 hPa)
  • Melting Pt:149…153 °C
  • Density:1,49 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:200-287-4

 

Specifications

Specification Test Results

  • Appearance:
    White to yellowish-white to greyish-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    149 to 153 °C
  • Loss on drying:
    ≤0.5 % (105 °C)
  • UV spectrophotometry:
    98.0 to 102.0 % (on dried substance)
  • Specific optical rotation:
    +18.5° to +20.5° (20 °C, 589 nm) (c=6, ethanol)

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.