X-Gal 99%, pure
Glass bottle, 2,5 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
- CAS nummer:7240-90-6
- Formel:C₁₄H₁₅BrClNO₆
- MDL nummer:MFCD00005666
- Molecular Weight:408.63 g/mol
- Smeltende Pt:230 °C (dec.)
- Opbevaringstemperatur:Køleskab
- EINECS:230-640-8
Specifikationer
Specifikationstestresultater
- Appearance:White Powder
- Infrared spectrum:Conforms
- Water:≤1 % (K.F.)
- HPLC:≥98.5 %
- Specific optical rotation:-60° to -64° (20°C, 589 nm) (c=1, H2O/DMF 1/1)
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



