Borsyre 99.5% for analysis
Plastic drum, 10 kg
Leverandør: Thermo Fisher Scientific
Borsyre 99.5% for analysis
Katalog # ACRO217080100
Leverandør: Thermo Fisher Scientific
CAS nummer: 10043-35-3
Gå til:
Product Details & Documents
Synonymer: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
- CAS nummer:10043-35-3
- Formel:B(OH)₃
- MDL nummer:MFCD00011337
- Molecular Weight:61.83 g/mol
- Merck Index:13,01326
- Kogende Pt:185 °C (1013 hPa)
- Smeltende Pt:169 °C (dec.)
- Tæthed:1,44 g/cm³ (25 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:233-139-2
- NÅ:01-2119486683-25
Documents
ACRO21708
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder or crystals
- pH:3.6 to 4.0 (4 % at 20°C)
- Titration with NaOH:≥99.4 %
- Chloride (Cl):≤3 ppm
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤5 ppm
- Arsenic (As):≤0.5 ppm
- Cadmium (Cd):≤5 ppm
- Calcium (Ca):≤20 ppm
- Copper (Cu):≤5 ppm
- Iron (Fe):≤1 ppm
- Magnesium (Mg):≤5 ppm
- Zinc (Zn):≤5 ppm
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 % (in methanol)
- Non-volatiles w. MeOH/HCl:≤0.05 %
Specifikationer
- Pakning:Plastic drum
- Pakn:10 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



