Borsyre 99.5% for analysis
Plastic bottle, 2,5 kg
: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
- :10043-35-3
- :B(OH)₃
- :MFCD00011337
- :61.83 g/mol
- :13,01326
- :185 °C (1013 hPa)
- :169 °C (dec.)
- :1,44 g/cm³ (25 °C)
- :Stue temperatur
- :233-139-2
- :01-2119486683-25
- Appearance:White Crystalline powder or crystals
- pH:3.6 to 4.0 (4 % at 20°C)
- Titration with NaOH:≥99.4 %
- Chloride (Cl):≤3 ppm
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤5 ppm
- Arsenic (As):≤0.5 ppm
- Cadmium (Cd):≤5 ppm
- Calcium (Ca):≤20 ppm
- Copper (Cu):≤5 ppm
- Iron (Fe):≤1 ppm
- Magnesium (Mg):≤5 ppm
- Zinc (Zn):≤5 ppm
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 % (in methanol)
- Non-volatiles w. MeOH/HCl:≤0.05 %
- Pakning:Plastic bottle
- Pakn:2,5 kg



