2-Propanol 99.5% for analysis
Plastic drum, 10 L
Leverandør: Thermo Fisher Scientific
2-Propanol 99.5% for analysis
Katalog # ACRO184130100
Leverandør: Thermo Fisher Scientific
CAS nummer: 67-63-0
Gå til:
Product Details & Documents
Synonymer: Isopropylalkohol
- CAS nummer:67-63-0
- Formel:(CH₃)₂CHOH
- MDL nummer:MFCD00011674
- Molecular Weight:60.1 g/mol
- Kogende Pt:81…83 °C (760 mmHg)
- Smeltende Pt:–89,5 °C
- Tæthed:0,785 g/cm³
- Flash Pt:12 °C
- Opbevaringstemperatur:Stue temperatur
- FN:1219
- ADR:3,II
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Water:≤0.1 % (Coulometric)
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Acidity/alkalinity:≤0.0001 meq/g corresp. 0.0007% C3H6O2
- Substance darkened by H2SO4:≤10 APHA
- Cadmium (Cd):≤0.05 ppm
- Calcium (Ca):≤0.5 ppm
- Copper (Cu):≤0.05 ppm
- Iron (Fe):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Sodium (Na):≤1 ppm
- Sulfur (S):≤0.5 ppm
- Zinc (Zn):≤0.1 ppm
Specifikationer
- Pakning:Plastic drum
- Pakn:10 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



