Kaliumchlorid 99+% for analysis
Fibre drum, 25 kg
Leverandør: Thermo Fisher Scientific
Kaliumchlorid 99+% for analysis
Katalog # ACRO196770250
Leverandør: Thermo Fisher Scientific
CAS nummer: 7447-40-7
Gå til:
Product Details & Documents
Synonymer: Potassiumchloride
- CAS nummer:7447-40-7
- Formel:KCl
- MDL nummer:MFCD00011360
- Molecular Weight:74.55 g/mol
- Merck Index:14,07621
- Kogende Pt:1420 °C (760 mmHg)
- Smeltende Pt:770 °C
- Tæthed:1,98 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-211-8
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder or crystals
- Titration Argentometric:99.0 to 100.5 % (on dried substance)
- Loss on drying:≤0.5 % (1 g, 105°C, 3 h)
- Acidity/alkalinity:complies
- Bromide (Br):≤1000 ppm
- Iodide (I):≤50 ppm
- Sulfate (SO4):≤100 ppm
- Aluminium (Al):≤1 ppm
- Arsenic (As):≤0.5 ppm
- Cadmium (Cd):≤1 ppm
- Calcium (Ca):≤50 ppm
- Iron (Fe):≤10 ppm
- Lead (Pb):≤2 ppm
- Magnesium (Mg):≤50 ppm
- Sodium (Na):≤700 ppm
- Heavy metals (as Pb):≤5 ppm
Specifikationer
- Pakning:Fibre drum
- Pakn:25 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



