Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Colchicin 97%

Colchicin 97%

Glass bottle, 5 g
Katalog # ACRO227120050
Giftig
Sundhed
Leverandør:   Thermo Fisher Scientific
Colchicin 97%
Katalog # ACRO227120050
CAS nummer:  64-86-8
Colchicin 97%
Colchicin 97%

Glass bottle, 5 g

CAS nummer:  64-86-8
Price
3.410,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  7α-H-Colchicine, (S)-(-)-Colchicine
  • CAS nummer:64-86-8
  • Formel:C₂₂H₂₅NO₆
  • MDL nummer:MFCD00078484
  • Molecular Weight:399.44 g/mol
  • Merck Index:12,02536
  • Smeltende Pt:150…160 °C (dec.)
  • Tæthed:1,32 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-598-5
  • FN:1544
  • ADR:6.1,I

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Light yellow Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤2 % (K.F.)
  • HPLC:
    ≥96.0 % on anhydrous and solvent-free substance
  • Specific optical rotation:
    -240° to -250° (20°C, 589 nm) (c=1, alcohol) (on anhydrous and solvent-free substance

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    5 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.