Borsyre 99+%, extra pure
Plastic drum, 10 kg
Leverandør: Thermo Fisher Scientific
Borsyre 99+%, extra pure
Katalog # ACRO180570100
Leverandør: Thermo Fisher Scientific
CAS nummer: 10043-35-3
Gå til:
Product Details & Documents
Synonymer: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
- CAS nummer:10043-35-3
- Formel:B(OH)₃
- MDL nummer:MFCD00011337
- Molecular Weight:61.83 g/mol
- Merck Index:13,01326
- Kogende Pt:185 °C (1013 hPa)
- Smeltende Pt:169 °C (dec.)
- Tæthed:1,44 g/cm³ (25 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:233-139-2
- NÅ:01-2119486683-25
Documents
ACRO18057
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Powder or crystals or granules
- Titration with NaOH:≥99.0 %
- Chloride (Cl):≤10 ppm
- Sulfate (SO4):≤145 ppm
- Iron (Fe):≤10 ppm
Specifikationer
- Pakning:Plastic drum
- Pakn:10 kg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



