Natriummetabisulfit for analysis
Plastic bottle, 2,5 kg
Leverandør: Thermo Fisher Scientific
Natriummetabisulfit for analysis
Katalog # ACRO223070025
Leverandør: Thermo Fisher Scientific
CAS nummer: 7681-57-4
Gå til:
Product Details & Documents
Synonymer: Natriumdisulfit, di-Natriumdisulfit, Natriumpentaoxodisulfat
- CAS nummer:7681-57-4
- Formel:Na₂S₂O₅
- MDL nummer:MFCD00167602
- Molecular Weight:190.11 g/mol
- Merck Index:12,08731
- Smeltende Pt:150 °C (dec.)
- Tæthed:2,476 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-548-0
- ADR:8,III
Documents
ACRO22307
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder
- Titration Iodimetric:≥58.5 % (SO2)
- Arsenic (As):≤1 ppm
- Iron (Fe):≤20 ppm
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.005 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:2,5 kg
- Cat. No.:ACRO223070025
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



