Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Jod 99.5+% for analysis

Glass bottle, 500 g
Katalog # ACRO212495000
Irriterende
Sundhed
Miljø
Leverandør:   Thermo Fisher Scientific
Jod 99.5+% for analysis
Katalog # ACRO212495000
CAS nummer:  7553-56-2
Jod 99.5+% for analysis
Jod 99.5+% for analysis

Glass bottle, 500 g

CAS nummer:  7553-56-2
Price
2.300,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
  • CAS nummer:7553-56-2
  • Formel:I₂
  • MDL nummer:MFCD00011355
  • Molecular Weight:253.81 g/mol
  • Kogende Pt:185 °C (760 mmHg)
  • Smeltende Pt:113 °C
  • Tæthed:4,9 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • FN:3495
  • ADR:8,III
Specifikationer

Specifikationstestresultater

  • Appearance:
    Dark blue to black Glistening spheres
  • Titration Iodometric:
    99.5 to 100.5 %
  • Chlorine and Bromine (as Cl):
    ≤0.005 %
  • Calcium (Ca):
    ≤10 ppm
  • Copper (Cu):
    ≤5 ppm
  • Iron (Fe):
    ≤50 ppm
  • Lead (Pb):
    ≤5 ppm
  • Magnesium (Mg):
    ≤5 ppm
  • Potassium (K):
    ≤50 ppm
  • Sodium (Na):
    ≤50 ppm
  • Zinc (Zn):
    ≤5 ppm
  • Insoluble matter:
    Passes test

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 g

Specifikationer