Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Jod 99.5+% for analysis

Jod 99.5+% for analysis

Glass bottle, 100 g
Katalog # ACRO212491000
Irriterende
Sundhed
Miljø
Leverandør:   Thermo Fisher Scientific
Jod 99.5+% for analysis
Katalog # ACRO212491000
CAS nummer:  7553-56-2
Jod 99.5+% for analysis
Jod 99.5+% for analysis

Glass bottle, 100 g

CAS nummer:  7553-56-2
Price
680,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
  • CAS nummer:7553-56-2
  • Formel:I₂
  • MDL nummer:MFCD00011355
  • Molecular Weight:253.81 g/mol
  • Kogende Pt:185 °C (760 mmHg)
  • Smeltende Pt:113 °C
  • Tæthed:4,9 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • FN:3495
  • ADR:8,III

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Dark blue to black Glistening spheres
  • Titration Iodometric:
    99.5 to 100.5 %
  • Chlorine and Bromine (as Cl):
    ≤0.005 %
  • Calcium (Ca):
    ≤10 ppm
  • Copper (Cu):
    ≤5 ppm
  • Iron (Fe):
    ≤50 ppm
  • Lead (Pb):
    ≤5 ppm
  • Magnesium (Mg):
    ≤5 ppm
  • Potassium (K):
    ≤50 ppm
  • Sodium (Na):
    ≤50 ppm
  • Zinc (Zn):
    ≤5 ppm
  • Insoluble matter:
    Passes test

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.