Jod 99.5+% for analysis
Glass bottle, 25 g
Leverandør: Thermo Fisher Scientific
Jod 99.5+% for analysis
Katalog # ACRO212490250
Leverandør: Thermo Fisher Scientific
CAS nummer: 7553-56-2
Gå til:
Product Details & Documents
- CAS nummer:7553-56-2
- Formel:I₂
- MDL nummer:MFCD00011355
- Molecular Weight:253.81 g/mol
- Kogende Pt:185 °C (760 mmHg)
- Smeltende Pt:113 °C
- Tæthed:4,9 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- FN:3495
- ADR:8,III
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Dark blue to black Glistening spheres
- Titration Iodometric:99.5 to 100.5 %
- Chlorine and Bromine (as Cl):≤0.005 %
- Calcium (Ca):≤10 ppm
- Copper (Cu):≤5 ppm
- Iron (Fe):≤50 ppm
- Lead (Pb):≤5 ppm
- Magnesium (Mg):≤5 ppm
- Potassium (K):≤50 ppm
- Sodium (Na):≤50 ppm
- Zinc (Zn):≤5 ppm
- Insoluble matter:Passes test
Specifikationer
- Pakning:Glass bottle
- Pakn:25 g



