Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Basic Violet 3, pure indicator

Basic Violet 3, pure indicator

Glass bottle, 500 g
Catalog # ACRO212125000
Irritant
Health
Corrosive
Environment
Basic Violet 3, pure indicator
Catalog # ACRO212125000
CAS Number:  548-62-9
Basic Violet 3, pure indicator
Basic Violet 3, pure indicator

Glass bottle, 500 g

CAS Number:  548-62-9
Price
2.590,00 kr
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  4-[4,4'-Bis(dimethylamino)benzhydryliden]cyclohexa-2,5-dien-1-yliden]dimethylammoniumchlorid, CI 42555
  • CAS Number:548-62-9
  • Formula:C₂₅H₃₀ClN₃
  • MDL Number:MFCD00011750
  • Molecular Weight:407.99 g/mol
  • Merck Index:13,04406
  • Melting Pt:173 °C (dec.)
  • Density:1,19 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:208-953-6
  • UN:3077
  • ADR:9,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Dark green Crystalline powder
  • Infrared spectrum:
    Conforms
  • UV spectrum:
    Complies
  • Sensitivity:
    (as indicator) Passes test

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    500 g

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.