Eddikesyre 99.8% for analysis
Plastic drum, 5 L
Leverandør: Thermo Fisher Scientific
Eddikesyre 99.8% for analysis
Katalog # 222140050.
Leverandør: Thermo Fisher Scientific
CAS nummer: 64-19-7
Gå til:
Product Details & Documents
Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS nummer:64-19-7
- Formel:H₃CCOOH
- MDL nummer:MFCD00036152
- Molecular Weight:60.05 g/mol
- Kogende Pt:117…118 °C (760 mmHg)
- Smeltende Pt:16…16,5 °C
- Tæthed:1,048 g/cm³
- Flash Pt:40 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-580-7
- FN:2789
- ADR:8,II
- NÅ:01-2119475328-30
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Water:≤0.2 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Titration with NaOH:≥99.8 %
- Dilution test:Passes test
- Chloride (Cl):≤1 ppm
- Sulfate (SO4):≤1 ppm
- Arsenic (As):≤0.5 ppm
- Cadmium (Cd):≤0.1 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.2 ppm
- Nickel (Ni):≤0.1 ppm
- Heavy metals (as Pb):≤0.5 ppm
- Acetic anhydride (C4H6O3):≤0.01 %
- KMnO4-reducing substance:Passes test
- K2Cr2O7-reducing substance:Passes test
Specifikationer
- Pakning:Plastic drum
- Pakn:5 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



