Bariumnitrat ACS
Plastic bottle, 500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
- CAS nummer:10022-31-8
- Formel:Ba(NO₃)₂
- MDL nummer:MFCD00003442
- Molecular Weight:261.34 g/mol
- Smeltende Pt:592 °C
- Tæthed:~ 3,23 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- FN:1446
- ADR:5.1,II
Documents
ACRO19255
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Crystals
- Titration Complexometric:≥99.0 %
- pH:5.0 to 8.0 (5 % at 25.0°C)
- Chloride (Cl):≤5 ppm
- Calcium (Ca):≤0.05 %
- Iron (Fe):≤2 ppm
- Potassium (K):≤0.005 %
- Sodium (Na):≤0.005 %
- Strontium (Sr):≤0.1 %
- Heavy metals (ICP-OES):≤5 ppm
- Insoluble matter:≤0.01 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



