Bariumnitrat ACS
Plastic bottle, 2,5 kg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
- CAS nummer:10022-31-8
- Formel:Ba(NO₃)₂
- MDL nummer:MFCD00003442
- Molecular Weight:261.34 g/mol
- Smeltende Pt:592 °C
- Tæthed:~ 3,23 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- FN:1446
- ADR:5.1,II
Specifikationer
Specifikationstestresultater
- Appearance:White Crystals
- Titration Complexometric:≥99.0 %
- pH:5.0 to 8.0 (5 % at 25.0°C)
- Chloride (Cl):≤5 ppm
- Calcium (Ca):≤0.05 %
- Iron (Fe):≤2 ppm
- Potassium (K):≤0.005 %
- Sodium (Na):≤0.005 %
- Strontium (Sr):≤0.1 %
- Heavy metals (ICP-OES):≤5 ppm
- Insoluble matter:≤0.01 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:2,5 kg



