Ammoniumhydrogendifluorid 95%, extra pure
Plastic bottle, 250 g
Leverandør: Thermo Fisher Scientific
Ammoniumhydrogendifluorid 95%, extra pure
Katalog # ACRO208822500
Leverandør: Thermo Fisher Scientific
CAS nummer: 1341-49-7
Gå til:
Product Details & Documents
Synonymer: Ammonium bifluoride, Ammonium hydrogen fluoride
- CAS nummer:1341-49-7
- Formel:NH₄HF₂
- MDL nummer:MFCD00012018
- Molecular Weight:57.04 g/mol
- Merck Index:13,00498
- Smeltende Pt:125 °C
- Tæthed:0,8 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:215-676-4
- FN:1727
- ADR:8,II
Documents
ACRO20882
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Flakes or scales
- Water:≤0.5 % (K.F.)
- Titration with NaOH:≥94 %
- Sulfite (SO3):≤100 ppm
- Sulfate (SO4):≤2000 ppm
- Iron (Fe):≤500 ppm
- Heavy metals (as Pb):≤200 ppm
- Impurity:≤5 % (Ammonium fluoride (NH4F))
Specifikationer
- Pakning:Plastic bottle
- Pakn:250 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



