Glycin 99+% for analysis
Fibre drum, 25 kg
Leverandør: Thermo Fisher Scientific
Glycin 99+% for analysis
Katalog # ACRO220910250
Leverandør: Thermo Fisher Scientific
CAS nummer: 56-40-6
Gå til:
Product Details & Documents
Synonymer: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS nummer:56-40-6
- Formel:NH₂CH₂COOH
- MDL nummer:MFCD00008131
- Molecular Weight:75.07 g/mol
- Merck Index:13,04504
- Kogende Pt:181 °C (1013 hPa)
- Smeltende Pt:240 °C
- Tæthed:1,595 g/cm³
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-272-2
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.2 % (105°C, 2 h)
- Sulfated ash:≤0.1 %
- Titration with HClO4:≥99.0 % (on dried substance)
- Chloride (Cl):≤40 ppm
- Sulfate (SO4):≤50 ppm
- Ammonium (NH4):≤200 ppm
- Arsenic (As):≤1 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
- Hydrolyzable substance:Clear solution
- Residual solvents:Meets the requirements
Specifikationer
- Pakning:Fibre drum
- Pakn:25 kg



