Glycin 99+% for analysis
Plastic drum, 5 kg
: Aminoacetic acid, Glycocoll, H-Gly-OH
- :56-40-6
- :NH₂CH₂COOH
- :MFCD00008131
- :75.07 g/mol
- :13,04504
- :181 °C (1013 hPa)
- :240 °C
- :1,595 g/cm³
- :Stue temperatur
- :200-272-2
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.2 % (105°C, 2 h)
- Sulfated ash:≤0.1 %
- Titration with HClO4:≥99.0 % (on dried substance)
- Chloride (Cl):≤40 ppm
- Sulfate (SO4):≤50 ppm
- Ammonium (NH4):≤200 ppm
- Arsenic (As):≤1 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
- Hydrolyzable substance:Clear solution
- Residual solvents:Meets the requirements
- Pakning:Plastic drum
- Pakn:5 kg
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



