Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(±)-2-Butanol 99+% for analysis

(±)-2-Butanol 99+% for analysis

Glass bottle, 2,5 L
Katalog # ACRO220290025
Irriterende
Brandfarlig
Leverandør:   Thermo Fisher Scientific
(±)-2-Butanol 99+% for analysis
Katalog # ACRO220290025
CAS nummer:  78-92-2
(±)-2-Butanol 99+% for analysis
(±)-2-Butanol 99+% for analysis

Glass bottle, 2,5 L

CAS nummer:  78-92-2
Price
1.110,00 kr
Log ind for tilgængelighed
1 * 2,5 L
Product Details & Documents
Synonymer:  (±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol
  • CAS nummer:78-92-2
  • Formel:H₃CCH₂CH(OH)CH₃
  • MDL nummer:MFCD00004569
  • Molecular Weight:74.12 g/mol
  • Merck Index:13,01540
  • Kogende Pt:99 °C (760 mmHg)
  • Smeltende Pt:–115 °C
  • Tæthed:0,81 g/cm³
  • Flash Pt:24 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-158-5
  • FN:1120
  • ADR:3,III

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Clear liquid
  • Water:
    ≤0.2 %
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Non-volatile matter:
    ≤0.001 %
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤0.5 ppm
  • Lead (Pb):
    ≤0.1 ppm
  • Free acid (C4H8O2):
    ≤0.003 %
  • 2-Propanol:
    ≤0.2 % (GC)

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2,5 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.