Caprinsyre 99%
Glass bottle, 1 kg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Decylic acid, Decanoic acid
- CAS nummer:334-48-5
- Formel:C₁₀H₂₀O₂
- MDL nummer:MFCD00004441
- Molecular Weight:172.27 g/mol
- Merck Index:13,01764
- Kogende Pt:268…270 °C (760 mmHg)
- Smeltende Pt:30…32 °C
- Tæthed:0,89 g/cm³
- Flash Pt:150 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:206-376-4
Documents
ACRO16727
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Colorless or white to cream Crystals and/or chunks or fused solid or liquid
- Infrared spectrum:Conforms
- Melting point:29 to 34 °C
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:1 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



