Benzophenon 99%, pure
Plastic drum, 5 kg
Supplier: Thermo Fisher Scientific
Benzophenon 99%, pure
Catalog # ACRO105560050
Supplier: Thermo Fisher Scientific
CAS Number: 119-61-9
Jump to:
Product Details & Documents
Synonyms: Diphenylmethanone, Diphenyl ketone
- CAS Number:119-61-9
- Formula:C₁₃H₁₀O
- MDL Number:MFCD00003076
- Molecular Weight:182.22 g/mol
- Merck Index:13,01099
- Boiling Pt:305 °C (760 mmHg)
- Melting Pt:47…49 °C
- Density:1,085 g/cm³ (20 °C)
- Flash Pt:143 °C
- Storage Temperature:Stue temperatur
- EINECS:204-337-6
- ADR:9,III
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:47 to 49 °C
- GC:≥98.5 %
Specifications
- Pakning:Plastic drum
- Pakn:5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



