Benz[a]anthracen 99%
Glass bottle, 250 mg
: Benz(alpha)anthracen, 1,2-Benzanthracen
- :56-55-3
- :C₁₈H₁₂
- :MFCD00003599
- :228.29 g/mol
- :12,01089
- :437,6 °C (760 mmHg)
- :158…161 °C
- :1,245 g/cm³ (20 °C)
- :Stue temperatur
- :200-280-6
- :3077
- :9,III
- Appearance:Beige to yellow-green Crystalline powder
- Infrared spectrum:Conforms
- Melting point:158 to 161 °C
- GC:≥98.5 %
- Optimum lasing:518 nm (in Methanol)
- Suitability:(for use as a laser dye) Passes test
- Molar absorption coefficient:19900 l/(mol.cm) (at Lambda max)
- Pakning:Glass bottle
- Pakn:250 mg



