L(+)-Arginin 98+%
Plastic bottle, 2,5 kg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH
- CAS nummer:74-79-3
- Formel:C₆H₁₄N₄O₂
- MDL nummer:MFCD00002635
- Molecular Weight:174.2 g/mol
- Merck Index:13,00785
- Kogende Pt:368 °C (1013 hPa)
- Smeltende Pt:223…224 °C (dec.)
- Tæthed:1,3 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-811-1
Documents
ACRO10499
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White to almost white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (1 g, 105°C)
- Residue after ignition:≤0.3 %
- Specific optical rotation:+25.8° to +28.5° (20°C, 589 nm) (c=8, 6 N HCl) (on dried substance)
- Titration with HClO4:98.0 to 102.0 % (on dried substance)
- Chloride (Cl):≤500 ppm
- Sulfate (SO4):≤300 ppm
- Iron (Fe):≤30 ppm
- Heavy metals (as Pb):≤15 ppm
Specifikationer
- Pakning:Plastic bottle
- Pakn:2,5 kg



