D-(+)-Cellobiose 98%
Glass bottle, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
- CAS Number:528-50-7
- Formula:C₁₂H₂₂O₁₁
- MDL Number:MFCD00136034
- Molecular Weight:342.30 g/mol
- Melting Pt:239 °C (dec.)
- Storage Temperature:Stue temperatur
- EINECS:208-436-5
Documents
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤0.5 % (K.F.)
- Solubility:(5 % in water) Clear colorless
- HPLC:≥97.5 % (ELSD)
- Specific optical rotation:+33° to +35° (20°C, 589 nm) (c=1, water) (after 15 h)
Specifications
- Pakning:Glass bottle
- Pakn:500 g



