Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
D-(+)-Cellobiose 98%

D-(+)-Cellobiose 98%

Glass bottle, 100 g
Katalog # ACRO108461000
Leverandør:   Thermo Fisher Scientific
D-(+)-Cellobiose 98%
Katalog # ACRO108461000
CAS nummer:  528-50-7
D-(+)-Cellobiose 98%
D-(+)-Cellobiose 98%

Glass bottle, 100 g

CAS nummer:  528-50-7
Price
3.140,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
  • CAS nummer:528-50-7
  • Formel:C₁₂H₂₂O₁₁
  • MDL nummer:MFCD00136034
  • Molecular Weight:342.30 g/mol
  • Smeltende Pt:239 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:208-436-5

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.5 % (K.F.)
  • Solubility:
    (5 % in water) Clear colorless
  • HPLC:
    ≥97.5 % (ELSD)
  • Specific optical rotation:
    +33° to +35° (20°C, 589 nm) (c=1, water) (after 15 h)

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    100 g
  • Cat. No.:
    ACRO108461000

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.