n-Nonadecan 99%
Glass bottle, 100 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: N-nonadecane
- CAS nummer:629-92-5
- Formel:H₃C(CH₂)₁₇CH₃
- MDL nummer:MFCD00009012
- Molecular Weight:268.53 g/mol
- Kogende Pt:330 °C (760 mmHg)
- Smeltende Pt:32…34 °C
- Tæthed:0,7845 g/cm³ (20 °C)
- Flash Pt:168 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:211-116-8
Documents
ACRO12910
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline solid
- Infrared spectrum:Conforms
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:100 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



