1-Naphtol 99+%
Plastic bottle, 2,5 kg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: α-Naphtol
- CAS nummer:90-15-3
- Formel:C₁₀H₈O
- MDL nummer:MFCD00003930
- Molecular Weight:144.17 g/mol
- Merck Index:13,06409
- Kogende Pt:278…280 °C (760 mmHg)
- Smeltende Pt:95…97 °C
- Tæthed:1,224 g/cm³ (20 °C)
- Flash Pt:125 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-969-4
- FN:2811
- ADR:6.1,III
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Beige to grey to brown Crystalline powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:95 to 97 °C
- GC:≥99.0 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:2,5 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



