Acetanilid 99+%
Plastic drum, 5 kg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: N-Phenylacetamide
- CAS nummer:103-84-4
- Formel:C₈H₉NO
- MDL nummer:MFCD00008674
- Molecular Weight:135.17 g/mol
- Merck Index:13,00051
- Kogende Pt:304 °C (760 mmHg)
- Smeltende Pt:111…115 °C
- Tæthed:1,22 g/cm³ (20 °C)
- Flash Pt:173 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-150-7
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White to beige to grey to light brown Powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:111 to 115 °C
- GC:≥99.0 %
Specifikationer
- Pakning:Plastic drum
- Pakn:5 kg



