(S)-(-)-Æblesyre 99%
Plastic bottle, 500 g
Leverandør: Thermo Fisher Scientific
(S)-(-)-Æblesyre 99%
Katalog # ACRO150595000
Leverandør: Thermo Fisher Scientific
CAS nummer: 97-67-6
Gå til:
Product Details & Documents
Synonymer: L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
- CAS nummer:97-67-6
- Formel:C₄H₆O₅
- MDL nummer:MFCD00064213
- Molecular Weight:134.09 g/mol
- Merck Index:13,05730
- Kogende Pt:322 °C (1013 hPa)
- Smeltende Pt:99…107 °C
- Tæthed:1,595 g/cm³ (20 °C)
- Flash Pt:220 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-601-5
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder or granules
- Infrared spectrum:Conforms
- Melting point:99 to 107 °C
- Sulfated ash:≤0.2 %
- Specific optical rotation:-1.5° to -3° (20 °C, 589 nm) (c=8.5, water)
- Titration with NaOH:≥98.5 %
- Chloride (Cl):≤100 ppm
- Sulfate (SO4):≤300 ppm
- Arsenic (As):≤2 ppm
- Heavy metals (as Pb):≤20 ppm
- Fumaric acid:≤0.2 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:500 g
- Cat. No.:ACRO150595000
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



