Pentylacetat 99%
Glass bottle, 2,5 L
: n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate
- :628-63-7
- :C₇H₁₄O₂
- :MFCD00009500
- :130.19 g/mol
- :149 °C (760 mmHg)
- :–70,8 °C
- :0,87 g/cm³
- :24 °C
- :Stue temperatur
- :211-047-3
- :1104
- :3,III
Documents
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4010 to 1.4030 (20°C, 589 nm)
- GC:≥98.5 %
- TLC:≥97.5 %
- Specific optical rotation:-10.8° to -11.8° (20°C, 589 nm) (c=2 in DMF)
- Pakning:Glass bottle
- Pakn:2,5 L



