2-Mercaptoethanol 99%, pure
Glass bottle, 2,5 L
Leverandør: Thermo Fisher Scientific
2-Mercaptoethanol 99%, pure
Katalog # ACRO125470025
Leverandør: Thermo Fisher Scientific
CAS nummer: 60-24-2
Gå til:
Product Details & Documents
Synonymer: 2-Hydroxyethyl mercaptan, β-Mercaptoethanol
- CAS nummer:60-24-2
- Formel:HSCH₂CH₂OH
- MDL nummer:MFCD00004890
- Molecular Weight:78.14 g/mol
- Merck Index:13,05894
- Kogende Pt:157…158 °C (760 mmHg)
- Smeltende Pt:–100 °C
- Tæthed:1,11 g/cm³
- Flash Pt:73 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-464-6
- FN:2966
- ADR:6.1,II
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4990 to 1.5020 (20°C, 589 nm)
- Water:≤0.5 %
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L



