5,5'-Dithiobis(2-nitrobenzosyre) 99%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
5,5'-Dithiobis(2-nitrobenzosyre) 99%
Catalog # ACRO117540250
Supplier: Thermo Fisher Scientific
CAS Number: 69-78-3
Jump to:
Product Details & Documents
Synonyms: DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
- CAS Number:69-78-3
- Formula:C₁₄H₈N₂O₈S₂
- MDL Number:MFCD00007140
- Molecular Weight:396.36 g/mol
- Melting Pt:240…245 °C (dec.)
- Storage Temperature:Køleskab
- EINECS:200-714-4
Documents
Specifications
Specification Test Results
- Appearance:Off-white to yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:240 to 245 °C
- Loss on drying:≤0.5 %
- Titration with NaOH:≥98.5 %
- Sulfate (SO4):≤0.005 %
- Ammonium (NH4):≤0.003 %
- Barium (Ba):≤0.005 %
- Calcium (Ca):≤0.01 %
- Iron (Fe):≤5 ppm
- Manganese (Mn):≤5 ppm
- Potassium (K):≤0.005 %
- Sodium (Na):≤0.005 %
- Strontium (Sr):≤0.005 %
- Heavy metals (ICP-OES):≤5 ppm
- Insoluble matter:≤0.005 %
Specifications
- Pakning:Glass bottle
- Pakn:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



