5,5'-Dithiobis(2-nitrobenzosyre) 99%
Glass bottle, 1 g
Leverandør: Thermo Fisher Scientific
5,5'-Dithiobis(2-nitrobenzosyre) 99%
Katalog # ACRO117540010
Leverandør: Thermo Fisher Scientific
CAS nummer: 69-78-3
Gå til:
Product Details & Documents
Synonymer: DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
- CAS nummer:69-78-3
- Formel:C₁₄H₈N₂O₈S₂
- MDL nummer:MFCD00007140
- Molecular Weight:396.36 g/mol
- Smeltende Pt:240…245 °C (dec.)
- Opbevaringstemperatur:Køleskab
- EINECS:200-714-4
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Off-white to yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:240 to 245 °C
- Loss on drying:≤0.5 %
- Titration with NaOH:≥98.5 %
- Sulfate (SO4):≤0.005 %
- Ammonium (NH4):≤0.003 %
- Barium (Ba):≤0.005 %
- Calcium (Ca):≤0.01 %
- Iron (Fe):≤5 ppm
- Manganese (Mn):≤5 ppm
- Potassium (K):≤0.005 %
- Sodium (Na):≤0.005 %
- Strontium (Sr):≤0.005 %
- Heavy metals (ICP-OES):≤5 ppm
- Insoluble matter:≤0.005 %
Specifikationer
- Pakning:Glass bottle
- Pakn:1 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



