Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2-Hexanon 98%

2-Hexanon 98%

Glass bottle, 2,5 L
Katalog # 146880025.
Irriterende
Sundhed
Brandfarlig
Leverandør:   Thermo Fisher Scientific
2-Hexanon 98%
Katalog # 146880025.
CAS nummer:  591-78-6
2-Hexanon 98%
2-Hexanon 98%

Glass bottle, 2,5 L

CAS nummer:  591-78-6
Price
9.600,00 kr
Log ind for tilgængelighed
1 * 2,5 L
Product Details & Documents
Synonymer:  Butylmethylketon
  • CAS nummer:591-78-6
  • Formel:C₆H₁₂O
  • MDL nummer:MFCD00009482
  • Molecular Weight:100.16 g/mol
  • Merck Index:13,06060
  • Kogende Pt:127 °C (760 mmHg)
  • Smeltende Pt:–57 °C
  • Tæthed:0,81 g/cm³
  • Flash Pt:23 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:209-731-1
  • FN:1224
  • ADR:3,III

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3995 to 1.4015 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2,5 L

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.