Oxalsyre standard solution 0.1 N
Plastic bottle, 2,5 L
Leverandør: Thermo Fisher Scientific
Oxalsyre standard solution 0.1 N
Katalog # ACRO124280025
Leverandør: Thermo Fisher Scientific
CAS nummer: 144-62-7
Gå til:
Product Details & Documents
Synonymer: Ethanedioic acid
- CAS nummer:144-62-7
- Formel:C₂H₂O₄
- MDL nummer:MFCD00002573
- Molecular Weight:90.04 g/mol
- Tæthed:1 g/cm³
- Opbevaringstemperatur:Stue temperatur
- EINECS:205-634-3
- ADR:8,III
Documents
ACRO12428
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless Solution
- Strength:0.0998 to 0.1002 N (20 °C)
- Melting point:116 to 119 °C
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
Specifikationer
- Pakning:Plastic bottle
- Pakn:2,5 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



